Learn More
Thermo Scientific™ Fluorescein-D22 Coactivator Peptide, 100 μM
Descripción
LanthaScreen™ fluorescent nuclear receptor coregulator peptides contain known interaction motifs and are labeled with fluorescein. These peptides are matched and validated to complement the LanthaScreen™ TR-FRET Nuclear Receptor Coregulator Assays. Assays developed using these reagents enable primary or secondary screening of nuclear receptor agonists and/or antagonists.
How they work
Ligand binding to nuclear receptors causes conformational changes in the receptor, resulting in a cascade of events, including dissociation of repressor proteins, association of coactivator proteins, and assembly of pol II and other transcriptional factors for activation of target genes. TR-FRET based assays can be developed using the LanthaScreen™ panel of fluorescein-labeled coregulator peptides to investigate conformational changes of nuclear receptors upon ligand binding, either by determining the affinity of ligand-bound receptor for different coregulator peptides, or by identifying additional agonists or antagonists via displacement or recruitment of a specific coregulator peptide. In the LanthaScreen™ TR-FRET RXR beta Coactivator Assay, a terbium (Tb)-labeled anti-GST antibody is used to indirectly label GSTtagged RXR beta protein. An agonist (9-cis Retinoic Acid) added to the receptor which, upon ligand binding, causes a conformational change resulting in recruitment of the Fl-D22 coactivator peptide (Figure 1). The binding of the fluorescent coregulator peptide to RXR beta causes an increase in the TR-FRET emission ratio (Figure 2).
Especificaciones
Especificaciones
| Contenido y almacenamiento | Almacenar en congelador (de - 5 a - 30 °C). |
| Para utilizar con (aplicación) | Target & Lead Identification & Validation, Nuclear Receptor Biology, Nuclear Receptor Proteins, Biochemical Nuclear Receptor Assays, Nuclear Receptor Coregulator Interaction, Industrial & Applied Science, Pharma & Biopharma |
| Tipo de etiqueta | Colorantes clásicos |
| Cantidad | 100 μl |
| Estado normativo | RUO - research use only |
| Condiciones de envío | Hielo seco |