Learn More
Fludarabine, MedChemExpress
Código de producto. 16474217
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
5 mg
10 mg
50 mg
Tamaño de la unidad:
10 miligramos
5 miligramos
50 miligramos
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
16474217 50 mg 50 miligramos
16470768 5 mg 5 miligramos
16438338 10 mg 10 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Código de producto. 16474217 Proveedor Medchem Express N.º de proveedor HYB0069
MedChemExpress Fludarabine (NSC 118218) is a DNA synthesis inhibitor and a fluorinated purine analogue with antineoplastic activity in lymphoproliferative malignancies. Fludarabine inhibits the cytokine-induced activation of STAT1 and STAT1-dependent gene transcription in normal resting or activated lymphocytes.
Fludarabine (NSC 118218) is a DNA synthesis inhibitor and a fluorinated purine analogue with antineoplastic activity in lymphoproliferative malignancies. Fludarabine inhibits the cytokine-induced activation of STAT1 and STAT1-dependent gene transcription in normal resting or activated lymphocytes.
Especificaciones
Nombre del producto químico o material Fludarabine
CAS 21679-14-1
Color Blanco
Notas de grado de pureza Research
Fórmula molecular C10H12FN5O4
Cantidad 50 mg
Sinónimo F-ara-A NSC 118218
Información de solubilidad DMSO : 25 mg/mL (87.65 mM; Need ultrasonic)
SMILES NC1=NC(F)=NC2=C1N=CN2[C@@H]3O[C@H](CO)[C@@H](O)[C@@H]3O
Peso molecular (g/mol) 285.23
Formula Weight (peso de la fórmula) 285.23
Porcentaje de pureza 99.85%
Grado Research
Forma física Powder
Para utilizar con (aplicación) Cancer-Kinase/protease