Learn More
PeproTech FLT3LG Polyclonal Antibody, PeproTech®, Invitrogen™

Descripción
AA Sequence of recombinant protein: TQDCSFQHSP ISSDFAVKIR ELSDYLLQDY PVTVASNLQD EELCGGLWRL VLAQRWMERL KTVAGSKMQG LLERVNTEIH FVTKCAFQPP PSCLRFVQTN ISRLLQETSE QLVALKPWIT RQNFSRCLEL QCQPDSSTLP PPWSPRPLEA TAPTA. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human Flt3-Ligand. Anti-Human Flt3-Ligand-specific antibody was purified by affinity chromatography employing an immobilized Human Flt3-Ligand matrix. Sandwich ELISA: To detect Human Flt3-Ligand by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.5-2.0 μg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Anti-Human Flt3-Ligand (500-P42Bt) as a detection antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human Flt3-Ligand. Western Blot: To detect Human Flt3-Ligand by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant Human Flt3-Ligand is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.
Especificaciones
Especificaciones
| Antígeno | FLT3LG |
| Aplicaciones | ELISA, Western Blot |
| Clasificación | Polyclonal |
| Concentración | 0.1-1.0 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with no preservative |
| génica | FLT3LG |
| N.º de referencia del gen | P49771 |
| Alias de gen | FL; Flt 3 ligand; Flt3; Flt3 L; Flt3 ligand; Flt3l; Flt3lg; fms related receptor tyrosine kinase 3 ligand; fms related tyrosine kinase 3 ligand; FMS-like tyrosine kinase 3 ligand; fms-related tyrosine kinase 3; fms-related tyrosine kinase 3 ligand; H-FLT-3-L; Ly72L; M-FLT-3-L; NEWGENE_2322792; SL cytokine |
| Símbolos de los genes | FLT3LG |
| Mostrar más |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.