Learn More
Florfenicol, MedChemExpress
Código de producto. 16492266
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
100 mg
500 mg
Tamaño de la unidad:
100 miligramos
500 miligramos
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
16492266 500 mg 500 miligramos
16438746 100 mg 100 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 16492266 Proveedor Medchem Express N.º de proveedor HYB1374
MedChemExpress Florfenicol, a commonly used veterinary antibiotic, is currently indicated for the treatment of bovine respiratory disease, and also used in aquaculture for the control of enteric septicemia in catfish.
Florfenicol, a commonly used veterinary antibiotic, is currently indicated for the treatment of bovine respiratory disease, and also used in aquaculture for the control of enteric septicemia in catfish.
Especificaciones
Nombre del producto químico o material Florfenicol
Forma física Solid
CAS 73231-34-2
Color Off-White
Cantidad 500 mg
Notas de grado de pureza Research
Fórmula molecular C12H14Cl2FNO4S
Para utilizar con (aplicación) COVID-19-immunoregulation
Sinónimo (-)-Florfenicol SCH-25298
Información de solubilidad DMSO : 100 mg/mL (279.17 mM; Need ultrasonic)
SMILES O=C(N[C@H](CF)[C@H](O)C1=CC=C(S(=O)(C)=O)C=C1)C(Cl)Cl
Peso molecular (g/mol) 358.21
Formula Weight (peso de la fórmula) 358.21
Porcentaje de pureza 98.0%