Learn More
Fisogatinib, MedChemExpress
Código de producto. 16485777
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
5 mg
10 mg
50 mg
100 mg
Tamaño de la unidad:
10 miligramos
100 miligramos
5 miligramos
50 miligramos
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
16485777 5 mg 5 miligramos
16479606 50 mg 50 miligramos
16444346 100 mg 100 miligramos
16413048 10 mg 10 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Código de producto. 16485777 Proveedor Medchem Express N.º de proveedor HY100492
MedChemExpress Fisogatinib (BLU-554) is a potent, highly selective and orally active fibroblast growth factor receptor 4 (FGFR4) inhibitor with an IC50 of 5 nM. Fisogatinib has significant anti-tumor activity in models of hepatocellular carcinoma (HCC) that are dependent on FGFR4 signalling.
Fisogatinib (BLU-554) is a potent, highly selective and orally active fibroblast growth factor receptor 4 (FGFR4) inhibitor with an IC50 of 5 nM. Fisogatinib has significant anti-tumor activity in models of hepatocellular carcinoma (HCC) that are dependent on FGFR4 signalling.
Especificaciones
Nombre del producto químico o material Fisogatinib
CAS 1707289-21-1
Color Blanco
Notas de grado de pureza Research
Fórmula molecular C24H24Cl2N4O4
Cantidad 5 mg
Sinónimo BLU-554
Información de solubilidad DMSO : 100 mg/mL (198.66 mM; Need ultrasonic)
SMILES C=CC(N[C@@H]1[C@H](NC2=NC=C3C=C(C4=C(Cl)C(OC)=CC(OC)=C4Cl)C=CC3=N2)COCC1)=O
Peso molecular (g/mol) 503.38
Formula Weight (peso de la fórmula) 503.38
Porcentaje de pureza 98.82%
Grado Research
Forma física Solid
Para utilizar con (aplicación) Cancer-Kinase/protease