Learn More
Fipexide, MedChemExpress
Código de producto. 16468626
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
100 mg
Tamaño de la unidad:
100 miligramos
16468626 100 mg 100 miligramos
1 options
Código de producto. 16468626 Proveedor Medchem Express N.º de proveedor HYB1124
MedChemExpress Fipexide, a parachloro-phenossiacetic acid derivative, is a nootropic drug. Fipexide reduces striatal adenylate cyclase activity. Fipexide has positive effect on cognitive performance by dopaminergic neurotransmission. Fipexide is used for senile dementia research.
Fipexide, a parachloro-phenossiacetic acid derivative, is a nootropic drug. Fipexide reduces striatal adenylate cyclase activity. Fipexide has positive effect on cognitive performance by dopaminergic neurotransmission. Fipexide is used for senile dementia research.
Especificaciones
Nombre del producto químico o material Fipexide
CAS 34161-24-5
Color Blanco
Notas de grado de pureza Research
Fórmula molecular C20H21ClN2O4
Cantidad 100 mg
Información de solubilidad DMSO : 50 mg/mL (128.59 mM; Need ultrasonic)
SMILES ClC1=CC=C(OCC(N2CCN(CC3=CC=C(OCO4)C4=C3)CC2)=O)C=C1
Peso molecular (g/mol) 388.84
Formula Weight (peso de la fórmula) 388.84
Porcentaje de pureza 99.88%
Grado Research
Forma física Powder
Para utilizar con (aplicación) Neuroscience-Neuromodulation