Learn More
Tocris Bioscience™ Filipin III
Polyene antibiotic; inhibits formation of pathological isoform of prion protein
Marca: Tocris Bioscience™ 6250/1
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Filipin III is a polyene antibiotic. Binds sterols in fungal membranes. Also inhibits formation of the pathological form of prion protein (PrP-res) from the normal membrane-bound (PrP-sen) form. Also fluorescent cholesterol stain.Especificaciones
| Polyene Antibiotic, Inhibits Formation of Pathological Isoform of Prion Protein | |
| C35H58O11 | |
| Store at -20°C | |
| 1 mg |
| 480-49-9 | |
| 654.83g/mol | |
| Antibiotics |
Título del producto
For Research Use Only