Learn More
MedChemExpress Fibronectin Adhesion-promoting Peptide
Descripción
It promotes assembly of mesenchymal stem cell (MSC) spheroids into larger aggregates. Fibronectin Adhesion-promoting Peptide directly promotes the adhesion, spreading, and migration of endothelial cells by reacting with heparin binding domains of cells.
Especificaciones
Especificaciones
| Pureza | 99.13% |
| CAS | 125720-21-0 |
| Contenido y almacenamiento | Powder -80°C 2 years, -20°C 1 year∣In solvent -80°C 6 months, -20°C 1 month |
| Para utilizar con (aplicación) | Neuroscience-Neuromodulation |
| Peso molecular | 1023.19 g/mol |
| Tipo de producto | Peptides |
| Cantidad | 10 mg |
| Condiciones de envío | Room temperature |
| Solubilidad | H2O : 2 mg/mL (1.95 mM∣Need ultrasonic and warming) |
| Sinónimo | Heparin Binding Peptide |