missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FGFR2 Monoclonal Antibody (10), Invitrogen™

Descripción
Specificity: Human FGFR2/CD332
FGFR2 is a member of the FGFR family of receptor tyrosine kinases. This family is known to regulate a host of cellular functions including angiogenesis, mitogenesis, osteogenesis, myogenesis, carcinogenesis, cellular differentiation, and tissue repair after injury. The FGFR family has also been implicated in a number of diseases including cancer, rheumatoid arthritis, and diabetic retinopathy. FGFR family members differ from one another in their ligand affinities and tissue distribution. Mutations in this gene are associated with Crouzon syndrome, Pfeiffer syndrome, Craniosynostosis, Apert syndrome, Jackson-Weiss syndrome, Beare-Stevenson cutis gyrata syndrome, Saethre-Chotzen syndrome, and syndromic craniosynostosis. Multiple alternatively spliced transcript variants encoding different isoforms have been noted for this gene.
Especificaciones
Especificaciones
| Antígeno | FGFR2 |
| Aplicaciones | ELISA |
| Clasificación | Monoclonal |
| Clon | 10 |
| Concentración | 1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with no preservative |
| génica | FGFR2 |
| N.º de referencia del gen | P21802 |
| Alias de gen | AU043015; AW556123; bacteria-expressed kinase; BBDS; BEK; BEK fibroblast growth factor receptor; BFR-1; CD332; CEK3; CFD1; ECT1; FGF Receptor; FGF Receptor 2; FGF-10; FGFR; FGFR2; FGFR-2; Fgfr7; Fgfr-7; fibroblast growth factor receptor 2; JWS; Keratinocyte growth factor receptor; KGFR; KGFRTr; KSAM; K-SAM; protein tyrosine kinase, receptor like 14; Soluble KGFR; svs; TK14; TK25 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?