Learn More
Tocris Bioscience™ FFN 102 mesylate
Selective fluorescent substrate of DAT and VMAT2
Marca: Tocris Bioscience™ 5200/10
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
FFN 102 mesylate is a pH responsive fluorescent false neurotransmitter (FFN). Selective dopamine transporter (DAT) and VMAT2 substrate. Exhibits no significant binding to a panel of 38 CNS receptors, including dopamine and serotonin receptors. Inhibits dopamine uptake. Excitation maxima are 340 nm at pH 5 and 370 nm at pH 7.5. Emission maximum is 435 nm at both pH 5 and 7.5.Especificaciones
| >99% | |
| 1883548-92-2 | |
| 10 mg |
Título del producto
For Research Use Only