Learn More
Fexaramine, MedChemExpress
Código de producto. 16410617
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
10 mg
50 mg
Tamaño de la unidad:
10 miligramos
50 miligramos
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
16410617 50 mg 50 miligramos
16404288 10 mg 10 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 16410617 Proveedor Medchem Express N.º de proveedor HY10912
MedChemExpress Fexaramine is a potent and selective FXR agonist with an EC50 of 25 nM. Fexaramine has no activity against hRXRα, hPPARαγδ, mPXR, hPXR, hLXRα, hTRβ, hRARβ, mCAR, mERRγ, and hVDR receptors.
Fexaramine is a potent and selective FXR agonist with an EC50 of 25 nM. Fexaramine has no activity against hRXRα, hPPARαγδ, mPXR, hPXR, hLXRα, hTRβ, hRARβ, mCAR, mERRγ, and hVDR receptors.
Especificaciones
Nombre del producto químico o material Fexaramine
CAS 574013-66-4
Color Yellow
Notas de grado de pureza Research
Fórmula molecular C32H36N2O3
Cantidad 50 mg
Información de solubilidad DMSO : 50 mg/mL (100.68 mM; Need ultrasonic)
SMILES CN(C)C1=CC=C(C2=CC=C(CN(C3=CC(/C=C/C(OC)=O)=CC=C3)C(C4CCCCC4)=O)C=C2)C=C1
Peso molecular (g/mol) 496.64
Formula Weight (peso de la fórmula) 496.64
Porcentaje de pureza 99.23%
Grado Research
Forma física Solid