missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FER Polyclonal Antibody
GREENER_CHOICE

Código de producto. 17752634
Change view
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
100 microlitros
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
17752634 100 μL 100 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 17752634 Proveedor Invitrogen™ N.º de proveedor PA5121237

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody

Positive test controls include: HeLa, HT-1080, SW480, LO2, NIH/3T3, Mouse liver, Mouse heart, Rat brain. The target is usually found in the following locations: Cell junction, Cell membrane, Cell projection, Cytoplasm, Cytoplasmic side, Membrane, Nucleus, Peripheral membrane protein, cell cortex, cytoskeleton. Immunogen sequence: YDITLPLLLD SLQKMQEEMI KALKGIFDEY SQITSLVTEE IVNVHKEIQM SVEQIDPSTE YNNFIDVHRT TAAKEQEIEF DTSLLEENEN LQANEIMWNN LTAESLQVML KTLAEELMQT QQMLLNKEEA VLELEKRIEE SSETCEKKSD IVLLLSQKQA LEELKQSVQQ LRCTEAKFSA QKELLEQKVQ ENDGKEPPPV VNYEEDARSV TSMERKERLS KFESIRHSIA GIIRSPKSAL GSSALSDMIS I.

FER is a member of the FPS/FES family of non-receptor tyrosine kinases. It regulates cell-cell adhesion and mediates signaling from the cell surface to the cytoskeleton via growth factor receptors. FER is found in the cytoplasm and in the nucleus, where it was associated with the chromatin fraction. Its ubiquitous expression and its subcellular location indicate that it may be involved in key regulatory processes. The group IV cytoplasmic protein-tyrosine kinase Fer has been linked to cellular signaling responses to many different stimuli, including growth factors and cytokines. Data suggest a biological role for Fer in regulation of leukocyte recruitment during the innate immune response.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno FER
Aplicaciones ELISA, Western Blot
Clasificación Polyclonal
Concentración 0.73 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
génica FER
N.º de referencia del gen P09760, P16591, P70451
Alias de gen AV082135; C330004K01Rik; c-FER; EC 2.7.10.2; Feline encephalitis virus-related kinase FER; FER; fer (fms/fps related) protein kinase; fer (fms/fps related) protein kinase, testis specific 2; fer (fps/fes related) tyrosine kinase; FER tyrosine kinase; Fert; Fert2; Flk; Flk_retired; Fujinami poultry sarcoma/Feline sarcoma-related protein Fer; p47; p94-Fer; phosphoprotein NCP94; PPP1R74; protein phosphatase 1, regulatory subunit 74; proto-oncogene c-Fer; proto-oncogene tyrosine-protein kinase Fer; TYK3; Tyrosine kinase 3; tyrosine protein kinase FLK; tyrosine-protein kinase Fer; Tyrosine-protein kinase FLK
Símbolos de los genes FER
Especie del huésped Rabbit
Inmunógeno Recombinant fusion protein containing a sequence corresponding to amino acids 200-450 of human FER (NP_005237.2).
Método de purificación Affinity chromatography
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 14158, 2241, 301737
Especies diana Human, Mouse, Rat
Contenido y almacenamiento -20°C, Avoid Freeze/Thaw Cycles
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.