Learn More
Fendiline Hydrochloride, Tocris Bioscience™
Inhibits KRas localization to the plasma membrane; also L-type calcium channel blocker
Marca: Tocris Bioscience 6407/10
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Fendiline Hydrochloride is a K-Ras inhibitor. Selectively inhibits K-Ras localization to the plasma membrane (IC50 = 9.64 μM). Blocks proliferation of cancer cell lines expressing oncogenic mutant K-Ras. Also inhibits L-type calcium channels (IC50 = 17 μM).Especificaciones
| Ras GTPase Inhibitors | |
| 13636-18-5 | |
| >98% | |
| 10 mg |