missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FBXL10 Polyclonal Antibody

Descripción
Peptide sequence: LSFFKRCGNI CHIDLRYCKQ VTKEGCEQFI AEMSVSVQFG QVEEKLLQKL Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 79%.
This gene encodes a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of ubiquitin protein ligase complex called SCFs, which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene belongs to the Fbls class. Multiple alternatively spliced transcript variants have been found for this gene, but the full-length nature of some variants has not been determined.
Especificaciones
Especificaciones
| Antígeno | FBXL10 |
| Aplicaciones | Western Blot |
| Clasificación | Polyclonal |
| Concentración | 0.5 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 2% sucrose and 0.09% sodium azide |
| génica | KDM2B |
| N.º de referencia del gen | Q8NHM5 |
| Alias de gen | [Histone-H3]-lysine-36 demethylase 1B; Cxxc2; CXXC-type zinc finger protein 2; Fbl10; F-box and leucine-rich repeat protein 10; F-box protein containing leucine repeats 10; F-box protein FBL10; F-box/LRR-repeat protein 10; Fbxl10; JEMMA (Jumonji domain, EMSY-interactor, methyltransferase motif) protein; Jhdm1b; JmjC domain-containing histone demethylation protein 1B; jumonji C domain-containing histone demethylase 1B; Jumonji domain-containing EMSY-interactor methyltransferase motif protein; Kdm2b; Kiaa3014; lysine (K)-specific demethylase 2B; lysine demethylase 2B; lysine-specific demethylase 2B; NDY1; PCCX2; Protein JEMMA; Protein-containing CXXC domain 2 |
| Símbolos de los genes | KDM2B |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?