missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FABP5 Polyclonal Antibody
GREENER_CHOICE

Código de producto. 15577632
Change view
Click to view available options
Cantidad:
100 μg
Tamaño de la unidad:
100 microgramos
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
15577632 100 μg 100 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 15577632 Proveedor Invitrogen™ N.º de proveedor PA547143

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Goat Polyclonal Antibody

In Western blots, less than 5% cross-reactivity with recombinant mouse FABP4 and recombinant human (rh) FABP3 is observed and less than 1% cross-reactivity with recombinant rat (rr) FABP2 and rrFABP1 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.

FABP5 (fatty acid binding protein 5 (psoriasis-associated)) is a proteinthat in humans is encoded by theFABP5 gene, also known as PAFABP, EFABP, E-FABP, KFABP, PA-FABP, Epidermal-type fatty acid-binding protein, Fatty acid-binding protein 5, Psoriasis-associated fatty acid-binding protein homolog. It is a family of small, highly conserved, cytoplasmic proteins that bind long-chain fatty acids and other hydrophobic ligands. The PAFABP cDNA encodes a 135-amino acid protein with molecular weight 15,164. This gene encodes the fatty acid binding protein found in epidermal cells, and was first identified as being upregulated inpsoriasistissue. It is though that FABPs roles include fatty acid uptake, transport, and metabolism. Kaczocha et al. (2009) identifiedFABP5and FABP7 as cytosolic proteins that transport AEA from the plasma membrane to subcellular fatty acid amide hydrolase, where it is hydrolyzed and inactivated. FABP3 did not show this specific transport function.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno FABP5
Aplicaciones Immunohistochemistry (Paraffin), Western Blot
Clasificación Polyclonal
Concentración 0.2 mg/mL
Conjugado Unconjugated
Formulación PBS with 5% trehalose and No Preservative
génica Fabp5
N.º de referencia del gen P55053, Q01469, Q05816
Alias de gen C-FABP; Cutaneous fatty acid-binding protein; DA11; EFABP; E-FABP; E-FABPPAFABP; Epidermal-type fatty acid-binding protein; epithelial fatty acid-binding protein; Fabp5; Fabpe; fatty acid binding protein 5; fatty acid binding protein 5 (psoriasis-associated); fatty acid binding protein 5, epidermal; fatty acid-binding protein 5; Fatty acid-binding protein, epidermal; fatty acid-binding protein, epidermal; uncharacterized protein LOC16592; keratinocyte lipid-binding protein; KFABP; Klbp; Mal1; PAFABP; PA-FABP; PA-FABPepidermal; Psoriasis-associated fatty acid-binding protein homolog
Símbolos de los genes Fabp5
Especie del huésped Goat
Inmunógeno E. coli-derived recombinant mouse FABP5/E-FABP Met1-Gln135.
Método de purificación Antigen Affinity Chromatography
Cantidad 100 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 140868, 16592, 2171
Especies diana Human, Mouse, Rat
Contenido y almacenamiento -20°C, Avoid Freeze/Thaw Cycles
Tipo de producto Antibody
Formulario Lyophilized
Isotype IgG
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.