missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ F(ab)-Goat anti-Mouse IgG2b Recombinant Secondary Antibody, Alexa Fluor™ Plus 488

Código de producto. 31722864
Change view
Click to view available options
Cantidad:
250 μL
Tamaño de la unidad:
250 microlitros
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
31722864 250 μL 250 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 31722864 Proveedor Invitrogen™ N.º de proveedor SA510382AFP488

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

F(ab)-Goat Recombinant Monoclonal Secondary Antibody

F(ab)-Goat anti-Mouse IgG2b Recombinant Secondary Antibody, Alexa Fluor™ Plus 488 is designed to specifically detect the mouse IgG2b isotype showing no detectable reactivity with immunoglobulins from any other tested species. This Fab fragment antibody is expressed in vitro and purified as a recombinant construct. It is also engineered to deliver enhanced stability and sensitivity. The advantages of recombinant antibodies include-better specificity, animal origin-free formulation, and lot-to-lot consistency. F(ab) fragments offer several advantages over full-length IgGs due to their smaller molecular size (~50 kDa vs. ~150 kDa), such as improved tissue penetration. The absence of the Fc region reduces nonspecific binding and background staining in Fc receptor-expressing cells and tissues, while also enabling increased flexibility in multiplexing assays. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno Mouse IgG2b
Aplicaciones Immunohistochemistry (Paraffin), Immunocytochemistry
Clasificación Recombinant Monoclonal
Clon 27H27L7
Concentración 1 mg/mL
Conjugado Alexa Fluor Plus 488
Formulación Proprietary buffer with 0.008% Bromonitrodioxane, 0.008% Methylisothiazolone; pH 6.3-7.3
Alias de gen IgG2; Immunoglobulin G; Immunoglobulin G2; ImmunoglobulinG; ImmunoglobulinG2
Especie del huésped Goat
Inmunógeno Purified Mouse IgG.
Método de purificación Affinity chromatography
Cantidad 250 μL
Estado normativo RUO
Primario o secundario Secondary
Especies diana Mouse
Contenido y almacenamiento 4°C, store in dark, DO NOT FREEZE!
Tipo de producto Secondary Antibody
Formulario Liquid
Isotype IgG F(ab)
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.