Learn More
Fisherbrand™ Termómetros sin mercurio Ertco™ Exact-Temp™
Código de producto. 11792183
Encompass_Preferred
Change view
Click to view available options
Intervalo de temperatura (métrico):
De -30 °C a +0 °C
De 20 °C a 130 °C
Para utilizar con (equipo):
Congeladores
Estufas
Tamaño de la unidad:
cada uno
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Intervalo de temperatura (métrico) Para utilizar con (equipo) unitSize
11792183 De -30 °C a +0 °C Congeladores cada uno
11712193 De 20 °C a 130 °C Estufas cada uno
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 11792183 Proveedor Fisherbrand™ N.º de proveedor F0101S
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Ver productos alternativos
Features an organic (mercury-free) filling. Ertco™ Exact-Temp™ Nonmercury Thermometers are high precision conventional bottle thermometers mounted in an unbreakable, filled plastic bottle for use in monitoring the temperature of freezers, refrigerators, incubators or ovens.
High-precision instrument to monitor temperature chambers, mounted in a shatterproof plastic bottle filled with liquid to ensure accurate readings.
  • Traceable to international standards (with certificate of conformance and accuracy)
  • Conventional thermometer works independent of any mains or battery connection
  • For accurate measurements, even with opening and closing of the appliance door
  • With engraved stems and thoroughly annealed for accuracy
  • Organic, mercury-free filling colored red for easy readings
  • Coated with PTFE resin to absorb shock and to contain glass and liquid in case of damage
  • With individual serial numbers
  • With double magnet for wall or door mounting
Certifications
A certificate is provided to indicate traceability to standards provided by NIST™
Especificaciones
Relleno del frasco Ethylene Glycol
Tamaño del frasco 30 mL
Para utilizar con (aplicación) Temperature Monitoring
Longitud total (imperial) 4.92 in.
Longitud total (métrico) 130 mm
Material Glass, PETG
Relleno del termómetro Líquido orgánico rojo
Tipo de producto Bottle Thermometer
Temperatura máxima 0 °C
Temperatura mín. (métrico) -30°C
Intervalo de temperatura (métrico) De -30 °C a +0 °C
Calibrated Sí
Para utilizar con (equipo) Congeladores
Exactitud ± 1 °C
Subdivisión 0,5 °C