Descripción
El control de la temperatura y la agitación de muestras son factores determinantes en muchos métodos analíticos UV-Visibles. Los accesorios termostáticos Peltier superan a las unidades calentador/enfriador tradicionales y ofrecen un rendimiento superior con un diseño compacto integrado, una configuración sencilla y un control de la temperatura preciso. Accesorios Thermo Scientific™ Peltier para espectrofotómetros Thermo Scientific™ Evolution™ serie 200 y 300 UV-Vis proporcionan termostatización y ajuste de la rampa con un preciso control de la temperatura a entre 0 y 110 °C.
Precise Thermostatting
Peltier Accessories for the Evolution UV-Visible Spectrophotometers utilize re-circulating water as an effective heat sink to allow the Peltier element to heat or cool the block over a wide temperature range. The single-cell holder can reach as low as 0 °C and as high at 110 °C. Thermo Scientific offers 3 Peltier options to cover many possible applications, including a multi-cell unit. Stirring control for the single cell holders is provided when you place a micro-stir bar inside the cuvette. Unlike thermostatting with a traditional re-circulating water bath where a large volume of liquid must be heated or cooled, heating or cooling the cell holder with the Peltier effect means more rapid and more precise temperature control. Choose between water or chemical coolant to best fit your needs.
Temperature Ramping
Thermo Scientific INSIGHT Pro Software allows you to control and monitor the block temperature of the Peltier unit and allows you to implement programmed temperature ramping with multiple ramp and hold cycles and variable temperature kinetics. Add a temperature probe hub and probes, and you can monitor the exact temperature in each cell as you perform heating and cooling cycles. The Insight Pro software also includes pre-set methods that can be used to study DNA denaturation and renaturation or other complex temperature programmed experiments.
Especificaciones
Especificaciones
| Tipo | Soporte de muestras multicelda |
| Para utilizar con (equipo) | Peltier rotatorio de 6 celdas |
| Incluye | Portaceldas Peltier, depósito de agua |