Learn More
Tocris Bioscience™ Evofosfamide
Descripción
Evofosfamide is a hypoxia-activated prodrug of the cytotoxin bromo-isophosphoramide mustard (Br-IPM). Evofosfamide is stable under normoxic conditions but is activated by cellular reductases in hypoxic conditions (IC50 values are 5.1 μM and 0.019 μM, in NSCLC cells under normoxic and hypoxic conditions, respectively). Br-IPM acts as a DNA alkylating agent, inducing intrastrand and interstrand crosslinks. Evofosfamide inhibits NEPC PDX tumor growth by 84.5% in adeno-CRPC and NEPC patient-derived xenograft models. When combined with gemcitabine (Cat. No. 3259), Evofosfamide inhibits primary tumor growth by 96% and significantly extends the survival time in the MIA PaCa-2 human pancreatic cancer orthotopic xenograft model.
Especificaciones
Especificaciones
| Nombre del producto químico o material | (1-Methyl-2-nitro-1H-imidazol-5-yl)methyl N,N'-bis(2-bromoethyl)phosphorodiamidate |
| CAS | 918633-87-1 |
| Cantidad | 10 mg |
| Objetivo | Apoptosis Inducer Compounds |
| Fórmula molecular | C9H16Br2N5O4P |
| Pureza | 0.98 |