Learn More
Thermo Scientific™ Evodiamine
Marca: Thermo Scientific™ J62771.MC
| Cantidad | 100 mg |
|---|
Descripción
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Especificaciones
| 100 mg | |
| -30°C to -10°C | |
| A vanilloid receptor agonist that induces apoptosis in leukemia U937 cells | |
| C19H17N3O |
| Off-white to pale yellow | |
| Solid | |
| 518-17-2 | |
| 303.40 |