missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ETNPPL Isoform 1 Polyclonal Antibody

Description
ETNPPL Isoform 1 Polyclonal Antibody for Western Blot, ICC/IF
Specifications
Specifications
| Antígeno | ETNPPL Isoform 1 |
| Aplicaciones | Western Blot, Immunocytochemistry |
| Clasificación | Polyclonal |
| Concentración | 0.1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| génica | ETNPPL |
| N.º de referencia del gen | Q8TBG4 |
| Alias de gen | 1300019H02Rik; Agxt2l1; AI195447; alanine--glyoxylate aminotransferase 2-like 1; alanine-glyoxylate aminotransferase 2-like 1; ethanolamine phosphate phospholyase; ethanolamine-phosphate phospho-lyase; ETNPPL |
| Símbolos de los genes | ETNPPL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?