missing translation for 'onlineSavingsMsg'
Learn More

Ethyl cycloheptylacetate, 98%, Thermo Scientific™

Product Code. 11495600
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
11495600 1 g 1g
11405610 5 g 5g
2 options
This item is not returnable. View return policy

Product Code. 11495600

Brand: Thermo Scientific Alfa Aesar H25866.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80246-70-4
Molecular Formula C11H20O2
Molecular Weight (g/mol) 184.279
MDL Number MFCD08437072
InChI Key WBZGWMNNCXEKSC-UHFFFAOYSA-N
Synonym ethyl cycloheptylacetate, cycloheptylacetic acid ethyl ester, cycloheptaneacetic acid, ethyl ester, ethyl cycloheptaneacetate, ethyl2-cycloheptylacetate, cycloheptyl-acetic acid ethyl ester
PubChem CID 14322005
IUPAC Name ethyl 2-cycloheptylacetate
SMILES CCOC(=O)CC1CCCCCC1

Specifications

Boiling Point 80°C to 85°C (2 mmHg)
Refractive Index 1.4525
Quantity 1 g
Formula Weight 184.28
Percent Purity 98%
Chemical Name or Material Ethyl cycloheptylacetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.