Learn More
Clorhidrato de etilo 5-aminovalerato, 96 %, Thermo Scientific Chemicals
Código de producto. 15408147
Cambiar vista
Click to view available options
Cantidad:
250 mg
1 g
Tamaño de la unidad:
1 g
250 miligramos
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
15408147 250 mg 250 miligramos
15498137 1 g 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 15408147 Proveedor Thermo Scientific Alfa Aesar N.º de proveedor H51682.MD
Solicitar formato a granel o personalizado
CAS: 29840-57-1 | C7H16ClNO2 | 181.66 g/mol
It is employed as intermediate for pharmaceutical.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Aplicaciones
Se emplea como intermediario en el sector farmacéutico.
Notas
Debe almacenarse en un lugar fresco. Mantenga el recipiente bien cerrado en un lugar seco y bien ventilado. Almacenar lejos de agentes oxidantes.
TRUSTED_SUSTAINABILITY
CAS 29840-57-1
Fórmula molecular C7H16ClNO2
Peso molecular (g/mol) 181.66
Número MDL MFCD12546205
Clave InChI JELXJZASXLHKDV-UHFFFAOYSA-N
Sinónimo ethyl 5-aminovalerate hydrochloride, ethyl 5-aminopentanoate hydrochloride, ethyl 5-aminovalerate hcl, 5-aminovaleric acid ethyl ester hydrochloride, pentanoic acid, 5-amino-, ethyl ester, hydrochloride, ethyl 5-aminopentanoate hcl, ethyl5-aminovaleratehydrochloride, ethyl 5-azanylpentanoate hydrochloride, 5-amino-valeric acid ethyl ester hydrochloride, 5-aminopentanoic acid ethyl ester hydrochloride
PubChem CID 16094861
Nombre IUPAC Etilo 5-aminopentanoato;clorhidrato
SMILES CCOC(=O)CCCCN.Cl
Especificaciones
Nombre del producto químico o material Ethyl 5-aminovalerate hydrochloride
Fórmula lineal H2N(CH2)4CO2CH2CH3·HCl
Cantidad 250 mg
Formula Weight (peso de la fórmula) 181.66
Porcentaje de pureza 96%