Learn More
Erlotinib, MedChemExpress
Código de producto. 16411936
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
100 mg
500 mg
Tamaño de la unidad:
100 miligramos
500 miligramos
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
16411936 500 mg 500 miligramos
16407896 100 mg 100 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 16411936 Proveedor Medchem Express N.º de proveedor HY50896
MedChemExpress Erlotinib (CP-358774) is a directly acting EGFR tyrosine kinase inhibitor, with an IC50 of 2 nM for human EGFR. Erlotinib reduces EGFR autophosphorylation in intact tumor cells with an IC50 of 20 nM. Erlotinib is used for the treatment of non-small cell lung cancer.
Erlotinib (CP-358774) is a directly acting EGFR tyrosine kinase inhibitor, with an IC50 of 2 nM for human EGFR. Erlotinib reduces EGFR autophosphorylation in intact tumor cells with an IC50 of 20 nM. Erlotinib is used for the treatment of non-small cell lung cancer.
Especificaciones
Nombre del producto químico o material Erlotinib
CAS 183321-74-6
Color Off-White
Notas de grado de pureza Research
Fórmula molecular C22H23N3O4
Cantidad 500 mg
Sinónimo CP-358774 NSC 718781 OSI-774
Información de solubilidad DMSO : 3.94 mg/mL (10.01 mM; ultrasonic and warming and heat to 40°C)
SMILES C#CC1=CC=CC(NC2=NC=NC3=C2C=C(C(OCCOC)=C3)OCCOC)=C1
Peso molecular (g/mol) 393.44
Formula Weight (peso de la fórmula) 393.44
Porcentaje de pureza 99.54%
Grado Research
Forma física Solid
Para utilizar con (aplicación) Cancer-Kinase/protease