Learn More
EPZ005687, MedChemExpress
Código de producto. 16496796
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
5 mg
10 mg
50 mg
100 mg
Tamaño de la unidad:
10 miligramos
100 miligramos
5 miligramos
50 miligramos
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
16496796 100 mg 100 miligramos
16496618 10 mg 10 miligramos
16459187 5 mg 5 miligramos
16402717 50 mg 50 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Código de producto. 16496796 Proveedor Medchem Express N.º de proveedor HY15555
MedChemExpress EPZ005687 is a potent and selective inhibitor of EZH2 with Ki of 24 nM, and has 50-fold selectivity against EZH1 and 500-fold selectivity against 15 other protein methyltransferases.
EPZ005687 is a potent and selective inhibitor of EZH2 with Ki of 24 nM, and has 50-fold selectivity against EZH1 and 500-fold selectivity against 15 other protein methyltransferases.
Especificaciones
Nombre del producto químico o material EPZ005687
Forma física Solid
CAS 1396772-26-1
Color Blanco
Cantidad 100 mg
Notas de grado de pureza Research
Fórmula molecular C32H37N5O3
Para utilizar con (aplicación) Cancer-programmed cell death
Información de solubilidad DMSO : 9.4 mg/mL (17.42 mM; Need ultrasonic and warming)
SMILES O=C(NCC1=C(C)C=C(C)NC1=O)C2=CC(C3=CC=C(CN4CCOCC4)C=C3)=CC5=C2C=NN5C6CCCC6
Peso molecular (g/mol) 539.67
Formula Weight (peso de la fórmula) 539.67
Porcentaje de pureza 95.11%