Learn More
PeproTech Eotaxin 2 Polyclonal Antibody, PeproTech®, Invitrogen™
Descripción
AA Sequence of recombinant protein: VTIPSSCCTS FISKKIPENR VVSYQLANGS ICPKAGVIFI TKKGHKICTD PKLLWVQRHI QKLDAKKNQP SKGAKAVRTK FAVQRRRGNS TEV. Preparation: Produced from sera of goats immunized with highly pure Recombinant Murine Eotaxin-2 (CCL24). Anti-Murine Eotaxin-2 (CCL24)-specific antibody was purified by affinity chromatography employing an immobilized Murine Eotaxin-2 (CCL24) matrix. Sandwich ELISA: To detect Murine Eotaxin-2 (CCL24) by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.5-2.0 μg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Anti-Murine Eotaxin-2 (CCL24) (500-P175GBt) as a detection antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Murine Eotaxin-2 (CCL24). Western Blot: To detect Murine Eotaxin-2 (CCL24) by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant Murine Eotaxin-2 (CCL24) is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.
Especificaciones
Especificaciones
| Antígeno | Eotaxin 2 |
| Aplicaciones | ELISA, Western Blot |
| Clasificación | Polyclonal |
| Concentración | 0.1-1.0 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with no preservative |
| génica | CCL24 |
| N.º de referencia del gen | Q9JKC0 |
| Alias de gen | CC chemokine CCL24; C-C motif chemokine 24; C-C motif chemokine ligand 24; Ccl24; chemokine (C-C motif) ligand 24; Ckb-6; CK-beta-6; Eosinophil chemotactic protein 2; Eotaxin2; eotaxin-2; eotaxin-2-like protein; MPIF2; MPIF-2; Myeloid progenitor inhibitory factor 2; Scya24; small inducible cytokine A24; small inducible cytokine subfamily A (Cys-Cys), member 24; small-inducible cytokine A24 |
| Símbolos de los genes | CCL24 |
| Mostrar más |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.