Learn More
Invitrogen™ γ-Glutamyl Transferase (GGT/γ-GT) Colorimetric Activity Kit
Descripción
The γ-Glutamyl Transferase (GGT/γ-GT) Colorimetric Activity Kit is a highly reliable and sensitive assay designed for the quantitative measurement of GGT in serum, plasma, and animal tissue samples. GGT is widely present in various organs of the human body, including the liver, kidney, and pancreas. GGT catalyzes the degradation of glutathione (GSH) and is involved in the regulation of glutathione levels in tissues, as well as the absorption and excretion of amino acids. Changes in GGT activity have been associated with various diseases, particularly those affecting the hepatobiliary system.
The assay utilizes the catalytic property of GGT to transfer a γ-glutamyl group from glutamyl p-nitroaniline to N-glycyl glycine, resulting in the production of p-nitroaniline. The p-nitroaniline has a characteristic absorption peak at 405 nm, allowing for the measurement of GGT activity.
Key features
• Simple and fast protocol—user-friendly, requiring minimal hands-on time and providing rapid results within a short incubation period
• Broad range of kits—across key analytes and targets within the most important metabolic pathways
• Consistent—strict quality control criteria to ensure reproducible and reliable results
Especificaciones
Especificaciones
| Intervalo del ensayo | 0.88–399.4 U/L |
| Sensibilidad del ensayo | 0.88 U/L |
| Contenido y almacenamiento | Store at 2–8°C. |
| Método de detección | Colorimétrico |
| Tipo de muestra | Plasma, Serum, Tissue Homogenates |
| Para utilizar con (equipo) | Lector de microplacas |
| Tiempo de incubación | 6 min. |
| CV interensayo | 6.2 |
| CV intraensayo | 4.2 |
| Cantidad | 96 pruebas |
| Mostrar más |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.