Learn More
Tocris Bioscience™ EHT 1864
GREENER_CHOICE
Código de producto. 18712494
Cambiar vista
Click to view available options
Cantidad:
10 mg
50 mg
Tamaño de la unidad:
10 miligramos
50 miligramos
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
18712494 10 mg 10 miligramos
18792724 50 mg 50 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 18712494 Proveedor Tocris Bioscience™ N.º de proveedor 3872/10
Potent Rac family inhibitor
Inhibitor of Rac family GTPases. Blocks activation by direct binding to Rac1, Rac1b, Rac2 and Rac3 (KD values are 40, 50, 60 and 250 nM respectively). Inhibits Rac, Ras and Tiam-induced growth transformation of NIH-3T3 fibroblasts. Reduces β-amyloid peptide production in vivo.
TRUSTED_SUSTAINABILITY
CAS 754240-09-0
Fórmula molecular C25H29Cl2F3N2O4S
Especificaciones
Nombre del producto químico o material Rho GTPase Inhibitors
Cantidad 10 mg
Formula Weight (peso de la fórmula) 581.47
Forma física Solid
Pureza >98%
Peso molecular 581.472