missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ EGFRvIII Recombinant Rabbit Monoclonal Antibody (RM419)
GREENER_CHOICE

Código de producto. 17156723 Tienda Thermo Scientific Productos
Change view
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
100 microlitros
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
17156723 100 μL 100 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 17156723 Proveedor Invitrogen™ N.º de proveedor MA536216

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Recombinant Monoclonal Antibody

MA5-36216 reacts to human Epidermal Growth Factor Receptor Variant III. No cross reactivity with wild type EGF receptor.

EGFRvIII is included in receptor tyrosine kinase binding ligands of the EGF family and activating several signaling cascades to convert extracellular cues into appropriate cellular responses. Known ligands include EGF, TGFA/TGF-alpha, amphiregulin, epigen/EPGN, BTC/betacellulin, epiregulin/EREG and HBEGF/heparin-binding EGF. Ligand binding triggers receptor homo- and/or heterodimerization and autophosphorylation on key cytoplasmic residues. The phosphorylated receptor recruits adapter proteins like GRB2 which in turn activates complex downstream signaling cascades. Activates at least 4 major downstream signaling cascades including the RAS-RAF-MEK-ERK, PI3 kinase-AKT, PLCgamma-PKC and STATs modules. May also activate the NF-kappa-B signaling cascade. Also directly phosphorylates other proteins like RGS16, activating its GTPase activity and probably coupling the EGF receptor signaling to the G protein-coupled receptor signaling. Also phosphorylates MUC1 and increases its interaction with SRC and CTNNB1/beta-catenin. Isoform 2 may act as an antagonist of EGF action.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno EGFRvIII
Aplicaciones ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Clasificación Recombinant Monoclonal
Clon RM419
Concentración 0.02 mg/mL
Conjugado Unconjugated
Formulación PBS with 1% BSA, 50% glycerol and 0.09% sodium azide
génica Egfr
N.º de referencia del gen P00533
Alias de gen 6720426O10Rik; 9030024J15Rik; A830025P08Rik; AI552599; avian erythroblastic leukemia viral (v-erbB) oncogene homolog; avian erythroblastic leukemia viral (v-erb-b) oncogene homolog; cell growth inhibiting protein 40; cell proliferation-inducing protein 61; Cntn5; CNTN6; contactin 5; contactin 6; contactin-5; Contactin-6; EGFR; EGFR-related peptide; epidermal growth factor receptor; Epidermal growth factor receptor formerly avian erythroblastic leukemia viral (v-erbB) oncogene homolog (Erbb1); epidermal growth factor receptor, formerly avian erythroblastic leukemia viral (v-erbB) oncogene homolog (Erbb1); Erbb; ERBB1; ErbB-1; erb-b2 receptor tyrosine kinase 1; Errb1; Errp; Gm507; HER1; hNB-2; HNB-2s; hNB-3; mENA; mNB-3; Nb-2; NB3; NB-3; neural adhesion molecule; neural recognition molecule NB-2; Neural recognition molecule NB-3; NISBD2; PIG61; Proto-oncogene c-ErbB-1; receptor tyrosine-protein kinase erbB-1; wa2; wa-2; Wa5; waved 2
Símbolos de los genes Egfr
Especie del huésped Rabbit
Inmunógeno A peptide corresponding to epidermal growth factor receptor variant III (EGFRvIII) (aa 1-100).
Método de purificación Protein A
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 1956
Especies diana Human
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.