missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Mouse EGF ELISA Development Kit (ABTS), PeproTech®
Descripción
EGF (Epidermal growth factor) exerts its actions by binding to the EGF Receptor, a 170 kDa protein kinase. Activation of EGFR initiates diverse cellular pathways in response to toxic environmental stimuli, or to EGF binding to the receptor, the EGFR forms homo- or heterodimers with other family members. Each dimeric receptor complex initiates a distinct signaling pathway by recruiting different Src homology 2 (SH2) containing effector proteins. EGF is far and wide expressed in kidney, cerebrum, prostrate and salivary glands. EGF acts as a potent mitogenic factor and the phosphorylated receptor recruits adapter proteins like GRB2 that activates complex downstream signaling cascades. EGF activates at least 4 major downstream signaling cascades including the RAS-RAF-MEK-ERK, PI3 kinase-AKT, PLCgamma-PKC and STAT modules. Research studies suggest the protein may also play important role in activating the NF-kappa-B signaling cascade. Defects in the EGGF gene are the cause of hypomagnesemia type 4 and dysregulation has been associated with the growth and progression of certain cancers.
Especificaciones
Especificaciones
| Número de acceso | P01132 |
| Intervalo del ensayo | Lot-specific |
| Sensibilidad del ensayo | Lot-specific |
| Conjugado | Biotina |
| Tipo de producto | ELISA |
| Para utilizar con (equipo) | Lector de microplacas colorimétrico |
| ID de gen (Entrez) | 13645 |
| Símbolo de gen | EGF |
| Contenido del kit | Capture Antibody, Detection Antibody, Protein Standard, HRP-conjugate |
| Etiqueta o tinte | HRP |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?