missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
Immunogen sequence: KEKFHKSQHW GFCNNVMMLV SDEKPGIGGE PLLGQKIKPK CSIYPKGDGS DVPSWVAFDK QVLSFDAYLE EEVLDKSQTN YRIR Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 80%.
EFHC2 (EF-hand domain-containing protein 2) is a gene located on the X chromosome at position Xp11.4. The protein encoded by EFHC2 is characterized by the presence of EF-hand motifs, which are helix-loop-helix structural domains capable of binding calcium ions. The EFHC2 protein is involved in various cellular processes, particularly those that are calcium-dependent, such as signal transduction and regulation of enzyme activities. EFHC2 has been implicated in neurological functions and conditions. For instance, it has been studied in the context of epilepsy, where variants in the gene are associated with increased susceptibility to juvenile myoclonic epilepsy. Additionally, EFHC2 is expressed in the brain and testis, suggesting roles in neurodevelopment and reproduction.
Especificaciones
Especificaciones
| Antígeno | EFHC2 |
| Aplicaciones | Immunohistochemistry (Paraffin), Western Blot |
| Clasificación | Polyclonal |
| Concentración | 0.6 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| génica | EFHC2 |
| N.º de referencia del gen | Q5JST6, Q9D485 |
| Alias de gen | 4933407D04Rik; dJ1158H2.1; EF-hand domain (C-terminal) containing 2; EF-hand domain containing 2; EF-hand domain-containing family member C2; Efhc2; mental retardation, X-linked 74; mRib72-2; MRX74 |
| Símbolos de los genes | EFHC2 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?
