Learn More
Tocris Bioscience™ Edaglitazone
Potent and selective PPARγ agonist; antidiabetic
Marca: Tocris Bioscience™ 4784/10
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Edaglitazone is a potent and selective PPARγ agonist (EC50 values are 35.6 and 1053 nM for PPARγ and PPARα cofactor recruitment respectively. Enhances insulin sensitivity in obese, but not lean, rats. Antidiabetic and orally bioavailable.Especificaciones
| Edaglitazone | |
| 5-[[4-[2-(5-Methyl-2-phenyl-4-oxazolyl)ethoxy]benzo[b]thien-7-yl]methyl]-2,4-thiazolidinedione | |
| Soluble to 100mM in DMSO | |
| Bioactive Small Molecule |
| 213411-83-7 | |
| 10 mg | |
| Solid |
Título del producto
Research Use Only. Not For Human or Veterinary Use