Learn More
STEMCELL Technologies EasySep™ Mouse Streptavidin RapidSpheres™ Isolation Kit
Código de producto. 17147981
Encompass
Cambiar vista
Click to view available options
Cantidad:
1 x 109 celdas
Tamaño de la unidad:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
17147981 1 x 109 celdas 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Código de producto. 17147981 Proveedor STEMCELL Technologies N.º de proveedor 19860
Please to purchase this item. Need a web account? Register with us today!
Immunomagnetic isolation of untouched mouse cells
  • The EasySep™ Mouse Streptavidin RapidSpheres™ Isolation Kit is designed for the depletion of single or multiple unwanted cell types labeled with biotinylated antibodies
  • Labeled cells from mouse splenocytes or other tissues are targeted for removal by Streptavidin RapidSpheres™ and separated without the use of columns using an EasySep™ magnet
  • Desired cells are untouched and poured off into a new tube
Especificaciones
Tipo de célula B Cells, Dendritic Cells, Granulocytes and Subsets, Hematopoietic Stem and Progenitor Cells, Macrophages, Marrow Stromal Cells, Mesenchymal Stem and Progenitor Cells, Monocytes, Mononuclear Cells, Myeloid Cells, NK Cells, Other, Plasma, T Cells
Para utilizar con (aplicación) Cell Isolation
Cantidad 1 x 109 celdas