Learn More
E7820, MedChemExpress
Código de producto. 16492237
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
5 mg
10 mg
25 mg
50 mg
Tamaño de la unidad:
10 miligramos
25 miligramos
5 miligramos
50 miligramos
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
16492237 50 mg 50 miligramos
16488697 5 mg 5 miligramos
16483426 25 mg 25 miligramos
16406108 10 mg 10 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Código de producto. 16492237 Proveedor Medchem Express N.º de proveedor HY14571
MedChemExpress E7820 (ER68203-00), an orally active aromatic sulfonamide derivative, is a unique angiogenesis inhibitor suppressing an expression of integrin alpha2 subunit on endothelium. E7820 inhibits rat aorta angiogenesis with an IC50 of 0.11 μg/ml. E7820 modulates α-1, α-2, α-3, and α-5 integrin mRNA expression. Antiangiogenic and antitumor activity.
E7820 (ER68203-00), an orally active aromatic sulfonamide derivative, is a unique angiogenesis inhibitor suppressing an expression of integrin alpha2 subunit on endothelium. E7820 inhibits rat aorta angiogenesis with an IC50 of 0.11 μg/ml. E7820 modulates α-1, α-2, α-3, and α-5 integrin mRNA expression. Antiangiogenic and antitumor activity.
Especificaciones
Nombre del producto químico o material E7820
CAS 289483-69-8
Color Light Yellow
Notas de grado de pureza Research
Fórmula molecular C17H12N4O2S
Cantidad 50 mg
Sinónimo ER68203-00
Información de solubilidad DMSO : 100 mg/mL (297.29 mM; Need ultrasonic)
SMILES O=S(C1=CC(C#N)=CC=C1)(NC2=C(NC=C3C#N)C3=C(C)C=C2)=O
Peso molecular (g/mol) 336.37
Formula Weight (peso de la fórmula) 336.37
Porcentaje de pureza 98.74%
Grado Research
Forma física Solid
Para utilizar con (aplicación) Cancer-programmed cell death