Learn More
Tocris Bioscience™ E 7820
Descripción
E 7820 is a α2 integrin inhibitor. Inhibits angiogenesis in vitro (IC50 = 0.11 - 0.25 μM). Also inhibits growth of human colorectal cancer cell lines. Enhances efficacy of erlotinib in a non-small-cell lung cancer xenograft model. Also acts as a molecular glue to induce proteosomal degradation of mRNA splicing factor RBM39.
Especificaciones
Especificaciones
| Nombre del producto químico o material | 3-Cyano-N-(3-cyano-4-methyl-1 H-indol-7-yl)benzenesulfonamide |
| CAS | 289483-69-8 |
| Cantidad | 10 mg |
| Objetivo | Integrin Receptor Inhibitors |
| Fórmula molecular | C17H12N4O2S |
| Formula Weight (peso de la fórmula) | 336.37 |
| Pureza | 98% |