Learn More
E 64c, MedChemExpress
Código de producto. 16485597
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
5 mg
10 mg
50 mg
Tamaño de la unidad:
10 miligramos
5 miligramos
50 miligramos
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
16485597 5 mg 5 miligramos
16462858 10 mg 10 miligramos
16419516 50 mg 50 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Código de producto. 16485597 Proveedor Medchem Express N.º de proveedor HY100227
MedChemExpress E 64c is a derivative of naturally occurring epoxide inhibitor of cysteine proteases, a Calcium-activated neutral protease (CANP) inhibitor and a very weak irreversible cathepsin C inhibitor. E 64c exhibits entry-blocking effect for MERS-CoV.
E 64c is a derivative of naturally occurring epoxide inhibitor of cysteine proteases, a Calcium-activated neutral protease (CANP) inhibitor and a very weak irreversible cathepsin C inhibitor. E 64c exhibits entry-blocking effect for MERS-CoV.
Especificaciones
Nombre del producto químico o material E 64c
Forma física Solid
CAS 76684-89-4
Color Blanco
Cantidad 5 mg
Notas de grado de pureza Research
Fórmula molecular C15H26N2O5
Para utilizar con (aplicación) Metabolism-protein/nucleotide metabolism
Información de solubilidad DMSO : 250 mg/mL (795.22 mM; Need ultrasonic)
SMILES O=C([C@H]1O[C@@H]1C(N[C@H](C(NCCC(C)C)=O)CC(C)C)=O)O
Peso molecular (g/mol) 314.38
Formula Weight (peso de la fórmula) 314.38
Porcentaje de pureza 99.73%