missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
Antibody detects endogenous levels of total DYNLRB1.
The DYNLRB1 gene is a member of the roadblock dynein light chain family. The encoded cytoplasmic protein is capable of binding intermediate chain proteins, interacts with transforming growth factor-beta, and has been implicated in the regulation of actin modulating proteins. Upregulation of this gene has been associated with hepatocellular carcinomas, suggesting that this gene may be involved in tumor progression. Alternative splicing results in multiple transcript variants. Pseudogenes of this gene have been defined on chromosomes 12 and 18.
Especificaciones
Especificaciones
| Antígeno | DYNLRB1 |
| Aplicaciones | Immunohistochemistry (Paraffin), Western Blot |
| Clasificación | Polyclonal |
| Concentración | 1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| génica | Dynlrb1 |
| N.º de referencia del gen | P62627, P62628, Q9NP97 |
| Alias de gen | 2010012N15Rik; 2010320M17Rik; 9430076K19Rik; AV124457; BITH; bithoraxoid-like protein; BLP; Dncl2a; DNLC2A; dynein light chain 2A, cytoplasmic; Dynein light chain roadblock-type 1; dynein, cytoplasmic, light chain 2A; dynein, cytoplasmic, light polypeptide 2A; dynein, light chain, roadblock-type 1; dynein-associated protein Km23; dynein-associated protein RKM23; DYNLRB1; HSPC162; hypothetical protein LOC513342; km23; km23-1; Roadblock domain-containing protein 1; ROBL/LC7-like 1; robl/LC7-like protein; ROBLD1; similar to dynein, cytoplasmic, light polypeptide 2A; zgc:56335 |
| Símbolos de los genes | Dynlrb1 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?
