Learn More
Tocris Bioscience™ Dynarrestin
Descripción
Dynarrestin is a dynein inhibitor. Inhibits hedgehog signaling in vitro (IC50 = 0.22 μM). Exhibits reversible inhibition of cytoplasmic dynein 1-dependent microtubule binding and cytoplasmic dynein 2-dependent intraflagellar transport. Inhibits tumor cell proliferation in a mouse medulloblastoma model.
Especificaciones
Especificaciones
| Nombre del producto químico o material | Ethyl 4-(4-(diethylamino)phenyl)-2-((2,4-difluorophenyl)amino)thiazole-5-carboxylate |
| CAS | 2222768-84-3 |
| Cantidad | 5 mg |
| Fórmula molecular | C22H23F2N3O2S |
| Formula Weight (peso de la fórmula) | 431.5 |
| Pureza | 98% |