Learn More
Tocris Bioscience™ DY 268
GREENER_CHOICE
Potent FXR antagonist
Marca:  Tocris Bioscience™ 5656/50
Encompass
Código de producto. 15787016
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Ver productos alternativos »
Descripción
Descripción
DY 268 is a potent FXR antagonist (IC50 = 7.5 nM). Exhibits no detectable cytotoxicity in cell-based assays.
Especificaciones
Especificaciones
1-[(3-Methoxyphenyl)methyl]-N-[4-methyl-3-(4-morpholinylsulfonyl)phenyl]-3-(4-methylphenyl)-1H-pyrazole-4-carboxamide
1609564-75-1