missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DRAK2 Polyclonal Antibody
GREENER_CHOICE

Código de producto. 11317113
Change view
Click to view available options
Cantidad:
100 μg
Tamaño de la unidad:
100 microgramos
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
11317113 100 μg 100 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 11317113 Proveedor Invitrogen™ N.º de proveedor PA519133

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Goat Polyclonal Antibody

This antibody is predicted to react with canine based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.

STK17B (also known as DRAK2) is a member of the serine/threonine kinase family and is related to death-associated protein kinase that triggers apoptosis. STK17B is selectively important for T-cell survival and inhibition of STK17B has therapeutic potential for autoimmune disease. T-cell survival depends on a balance of T-cell receptor and co-stimulatory signals and deficiency of STK17B can affect autoimmune disease susceptibility without generalized suppression of the immune system. Protein kinases play important roles in the signal transduction in response to a variety of external stimuli. Recently, several protein kinases have been identified that may also be involved in apoptotic process. Overexpression of a serine/threonine kinase, ZIP kinase can cause the morphological changes typical of apoptosis in NIH 3T3 cells. Sanjo et al, have identified two new protein kinases DRAK1 and DRAK2 (Death Receptor Associated Kinase 1 and 2). Both DRAKs and ZIP kinase share significant homology at the amino acid level. The kinase domains of both DRAKs, ZIP kinase are homologous to DAP (Death-associated protein) kinase, which is involved in the apoptotic signaling induced by interferon-g. Overexpression of DRAKs in NIH 3T3 cells lead to apoptosis. These transfection experiments suggest that C-terminal domain of DRAKs are important for induction of apoptosis.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno DRAK2
Aplicaciones Western Blot
Clasificación Polyclonal
Concentración 0.5 mg/mL
Conjugado Unconjugated
Formulación TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
génica STK17B
N.º de referencia del gen O94768
Alias de gen 3110009A03Rik; AI120141; DAP kinase-related apoptosis-inducing protein kinase 2; death-associated protein kinase-related 2; DRAK 2; DRAK2; serine/threonine kinase 17b; serine/threonine kinase 17b (apoptosis-inducing); serine/threonine-protein kinase 17B; ST17B; STK 17B; Stk17b
Símbolos de los genes STK17B
Especie del huésped Goat
Inmunógeno Synthetic peptide sequence (HSVRSSEDKTSK) corresponding to the internal amino acids of STK17B (aa 316-328).
Método de purificación Ammonium sulfate precipitation
Cantidad 100 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 9262
Especies diana Human
Contenido y almacenamiento -20°C, Avoid Freeze/Thaw Cycles
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.