Learn More
Tocris Bioscience™ DR 4485 Hydrochloride
Descripción
DR 4485 hydrochloride is a high affinity and selective 5-HT7 antagonist (pKi = 8.14). Exhibits selectivity for 5-HT7 over other 5-HT receptors. Inhibits 5-HT-induced cAMP accumulation in HEK-293 cells expressing the 5-HT7 receptor. Orally bioavailable.
Especificaciones
Especificaciones
| Nombre del producto químico o material | 6-Chloro-2a-[4-[4-(4-chlorophenyl)-3,6-dihydro-1(2H)-pyridinyl]butyl]-2a,3,4,5-tetrahydrobenz[cd]indol-2(1H)-one hydrochloride |
| CAS | 402942-53-4 |
| Cantidad | 10 mg |
| Fórmula molecular | C26H28Cl2N2O.HCl |
| Formula Weight (peso de la fórmula) | 491.88 |
| Pureza | 98% |