missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DPF1 Polyclonal Antibody
GREENER_CHOICE

Código de producto. 15709701 Tienda Thermo Scientific Productos
Change view
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
100 microlitros
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
15709701 100 μL 100 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 15709701 Proveedor Invitrogen™ N.º de proveedor PA561895

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody

Immunogen sequence: VLEALLCAET GEKKIELKEE ETIMDCQKQQ LLEFPHDLEV EDLEDDIPRR KNRAKGKAYG IGGLRKRQDT ASLEDR Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.

May have an important role in developing neurons by participating in regulation of cell survival, possibly as a neurospecific transcription factor. Belongs to the neuron-specific chromatin remodeling complex (nBAF complex). During neural development a switch from a stem/progenitor to a post-mitotic chromatin remodeling mechanism occurs as neurons exit the cell cycle and become committed to their adult state. The transition from proliferating neural stem/progenitor cells to post-mitotic neurons requires a switch in subunit composition of the npBAF and nBAF complexes. As neural progenitors exit mitosis and differentiate into neurons, npBAF complexes which contain ACTL6A/BAF53A and PHF10/BAF45A, are exchanged for homologous alternative ACTL6B/BAF53B and DPF1/BAF45B or DPF3/BAF45C subunits in neuron-specific complexes (nBAF). The npBAF complex is essential for the self-renewal/proliferative capacity of the multipotent neural stem cells. The nBAF complex along with CREST plays a role regulating the activity of genes essential for dendrite growth.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno DPF1
Aplicaciones Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Clasificación Polyclonal
Concentración 0.2 mg/mL
Conjugado Unconjugated
Formulación PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
génica DPF1
N.º de referencia del gen Q92782
Alias de gen Baf45b; BRG1-associated factor 45B; D4, zinc and double PHD fingers family 1; double PHD fingers 1; DPF1; NEU BAF45b; NEUD4; neuro-d4; neuro-d4 homolog; neuronal d4 domain family member; zinc finger protein neuro-d4
Símbolos de los genes DPF1
Especie del huésped Rabbit
Inmunógeno Recombinant protein corresponding to Human DPF1. Recombinant protein control fragment (Product #RP-101097).
Método de purificación Antigen affinity chromatography
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 8193
Especies diana Human
Contenido y almacenamiento Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.