Descripción
- Chattaway double spatula with a square end and a rounded, curved end for precise laboratory work
- Designed for ergonomic handling and accurate dosing in laboratory, medical diagnostic, and analytical environments
- Flat angular side enables precise portioning, swabbing, and cutting
- Flat rounded curved side allows gentle and efficient scraping, removal, and dispensing from round or hard-to-reach vessels
- Curved shape adapts to glass jars, reducing material loss and minimizing required effort
- Ergonomic bend supports a natural hand position during use
- Made from high-quality stainless steel for robustness, hygiene, and corrosion resistance
- Suitable for continuous use in demanding working environments
- Two functions: angular side for precise work and curved side for hard-to-reach areas
- Curved spatula angle enables ergonomic handling with minimized effort
- Gentle working in glass jars, curvature prevents scratches and adapts to vessel shape
- Efficient scraping along vessel walls helps reduce sample loss
Especificaciones
Especificaciones
| Esterilizables en autoclave | Autoclavable |
| Forma | Square |
| Tipo de producto | Double Spatula Chattaway |
| Longitud de la hoja (métrico) | 77/66 mm |
| Anchura (métrico) de la hoja | 11 mm |
| Material de la hoja | Stainless Steel |
| Material del mango | Stainless Steel |
| Material | Stainless Steel |
| Longitud (métrico) | 237 mm |