Learn More
Invitrogen™ Donkey anti-Rat IgG (H+L) Cross-Adsorbed Secondary Antibody, DyLight™ 650
Código de producto. 13407187
Change view
Click to view available options
Cantidad:
500 μg
Tamaño de la unidad:
500 microgramos
13407187 500 μg 500 microgramos
1 options
Código de producto. 13407187 Proveedor Invitrogen™ N.º de proveedor SA510029
Donkey Polyclonal Secondary Antibody
This antibody is cross-adsorbed and exhibits minimum reactivity to bovine, chicken, goat, human, mouse, rabbit and sheep.
Anti-Rat secondary antibodies are affinity-purified antibodies with well-characterized specificity for rat immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno Rat IgG (H+L) Cross-Adsorbed
Aplicaciones Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry
Clasificación Polyclonal
Concentración 0.5 mg/mL
Conjugado DyLight 650
Formulación PBS with 0.2% BSA and 0.09% sodium azide; pH 6.8 to 7.4
Especie del huésped Donkey
Inmunógeno Rat IgG-heavy and light chain.
Método de purificación Antigen affinity chromatography
Cantidad 500 μg
Estado normativo RUO
Primario o secundario Secondary
Especies diana Rat
Contenido y almacenamiento 4°C
Tipo de producto Secondary Antibody
Formulario Liquid
Isotype IgG