Learn More
Tocris Bioscience™ DMXAA
mSTING agonist; induces antitumor immunological responses
Marca: Tocris Bioscience™ 5601/50
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
DMXAA is a mSTING agonist; selective for mouse STING over human STING. Induces IFN-β and cytokine production from bone marrow-derived dendritic cells. Causes tumor regression following intratumoral administration in mouse tumor models. Induces an adaptive immune response to reduce systemic tumor growth and provides immunological memory against autologous tumor re-challenge. Also antiviral.Especificaciones
| 5,6-Dimethyl-9-oxo-9 H-xanthene-4-acetic acid | |
| 117570-53-3 | |
| 50 mg |