Learn More
Tocris Bioscience™ DMNB-caged-Serine
Descripción
DMNB-caged-Serine is a DMNB-caged serine. Photoactivated to release serine by visible blue light (405 nm) in cells. Incorporated into S.cerevisiae proteins when introduced into growth medium at millimolar concentrations.
Especificaciones
Especificaciones
| Nombre del producto químico o material | O-(4,5-Dimethoxy-2-nitrobenzyl)-L-serine |
| CAS | 780009-55-4 |
| Cantidad | 50 mg |
| Objetivo | Caged Compounds |
| Fórmula molecular | C12H16N2O7 |
| Pureza | 98% |