missing translation for 'onlineSavingsMsg'
Learn More

DLX4 Polyclonal Antibody, Invitrogen™

Código de producto. 11542422
Change view
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
100 microlitros
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
11542422 100 μL 100 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 11542422 Proveedor Invitrogen N.º de proveedor PA132425

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas
Ver productos alternativos

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody

PA1-32425 detects BP1 from human samples. PA1-32425 has been successfully used in immunohistochemistry, immunoprecipitation applications. The PA1-32425 immunogen is: Synthetic peptide corresponding to specifically to the human BP1 protein sequence (between residues 1-60). There is no homology with the peptide sequences used for immunogen of this antibody and the DLX4 or DLX7 proteins.

Many vertebrate homeo box-containing genes have been identified on the basis of their sequence similarity with Drosophila developmental genes. Members of the Dlx gene family contain a homeobox that is related to that of Distal-less (Dll), a gene expressed in the head and limbs of the developing fruit fly. The Distal-less (Dlx) family of genes comprises at least 6 different members, DLX1-DLX6. The DLX proteins are postulated to play a role in forebrain and craniofacial development. Three transcript variants have been described for this gene, however, the full length nature of one variant has not been described. Studies of the two splice variants revealed that one encoded isoform functions as a repressor of the beta-globin gene while the other isoform lacks that function.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno DLX4
Aplicaciones Immunohistochemistry, Immunoprecipitation, Western Blot
Clasificación Polyclonal
Concentración 1 mg/mL
Conjugado Unconjugated
génica DLX4
N.º de referencia del gen Q92988, P70436
Alias de gen BP1, DLX7, DLX8, DLX9
Símbolos de los genes DLX4
Especie del huésped Rabbit
Inmunógeno Synthetic peptide corresponding to specifically to the human BP1 protein sequence (between residues 1-60). There is no homology with the peptide sequences used for immunogen of this antibody and the DLX4 or DLX7 proteins.
Método de purificación Antigen affinity chromatography
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 13394, 1748
Especies diana Human, Mouse
Contenido y almacenamiento 4° C, do not freeze
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.