Learn More
DLK-IN-1, MedChemExpress
Código de producto. 16487557
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
5 mg
10 mg
25 mg
50 mg
100 mg
Tamaño de la unidad:
10 miligramos
100 miligramos
25 miligramos
5 miligramos
50 miligramos
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
16487557 5 mg 5 miligramos
16475606 100 mg 100 miligramos
16431077 50 mg 50 miligramos
16423256 25 mg 25 miligramos
16404818 10 mg 10 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Código de producto. 16487557 Proveedor Medchem Express N.º de proveedor HY114331
MedChemExpress DLK-IN-1 is a selective, orally active inhibitor of dual leucine zipper kinase (DLK, MAP3K12), with a Ki of 3 nM. DLK-IN-1 retains excellent CNS penetration and is well tolerated following multiple days of dosing at concentrations that exceed those required for DLK inhibition in the brain. DLK-IN-1 has activity in a model of Alzheimer’s Disease.
DLK-IN-1 is a selective, orally active inhibitor of dual leucine zipper kinase (DLK, MAP3K12), with a Ki of 3 nM. DLK-IN-1 retains excellent CNS penetration and is well tolerated following multiple days of dosing at concentrations that exceed those required for DLK inhibition in the brain. DLK-IN-1 has activity in a model of Alzheimer’s Disease.
Especificaciones
Nombre del producto químico o material DLK-IN-1
CAS 1620574-24-4
Color Blanco
Notas de grado de pureza Research
Fórmula molecular C20H24F3N5O2
Cantidad 5 mg
Información de solubilidad DMSO : 62.5 mg/mL (147.60 mM; Need ultrasonic)
SMILES CC(N1C([C@@H]2[C@]3([H])[C@@]2([H])CN(C4COC4)C3)=CC(C5=CN=C(N)C(OC(F)(F)F)=C5)=N1)C
Peso molecular (g/mol) 423.43
Formula Weight (peso de la fórmula) 423.43
Porcentaje de pureza 99.41%
Grado Research
Forma física Solid
Para utilizar con (aplicación) Neuroscience-Neurodegeneration