Learn More
Ácido DL-glutámico-2,3,3,4,4-d5, CDN
Product Code. 30319877
Change view
Click to view available options
Cantidad:
0.25 g
0.5 g
Unit Size:
0.25 g
0.50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cantidad unitSize
30319877 0.5 g 0.50 g
30319029 0.25 g 0.25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30319877 Supplier CDN Supplier No. CDND21930.5G
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Descubra más de 4000 compuestos deuterados de alta calidad, con envío directo desde stock para mayor rapidez. Ofrecemos muchas API únicas, incluidos ingredientes farmacéuticos activos, nitrosoaminas, aminoácidos, esteroides, gases, pesticidas y ácidos grasos.
TRUSTED_SUSTAINABILITY
Specifications
Nombre del producto químico o material DL-Glutamic Acid-d5
CAS 14341-79-8
Fórmula molecular C5 D5 H4 N O4
Cantidad 0.5 g
Sinónimo (±)-Glutamic Acid-d5, DL-Glu-d5, Glutaminic Acid-d5, NSC 206301-d5, NSC 9967-d5, DL-Glutamic-2,3,3,4,4-d5 Acid
Fórmula InChI InChI=1 S/C5H9NO4/c6-3(5(9)10)1-2-4(7)8/h3H,1-2,6H2,(H,7,8)(H,9,10)/i1D2,2D2,3 D
SMILES [2 H]C([2 H])(C(=O)O)C([2 H])([2 H])C([2 H])(N)C(=O)O
Nombre IUPAC ácido 2-amino-2,3,3,4,4-pentadeuteriopentanedioico
Peso molecular (g/mol) 152.16
Formula Weight (peso de la fórmula) 152.0845 g/mol
Porcentaje de pureza 98 atom % D, min 98% Chemical Purity