Learn More
DiZPK, MedChemExpress
Código de producto. 16481547
Encompass_Preferred
Change view
Click to view available options
Cantidad:
5 mg
10 mg
50 mg
Tamaño de la unidad:
10 miligramos
5 miligramos
50 miligramos
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
16481547 50 mg 50 miligramos
16465358 10 mg 10 miligramos
16438077 5 mg 5 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Código de producto. 16481547 Proveedor Medchem Express N.º de proveedor HY12801
MedChemExpress DiZPK is a photocrosslinker for identifying direct protein-protein interactions in living prokaryotic and eukaryotic cells.
DiZPK is a photocrosslinker for identifying direct protein-protein interactions in living prokaryotic and eukaryotic cells.
Especificaciones
Nombre del producto químico o material DiZPK
Forma física Solid
CAS 1337883-32-5
Color Blanco
Cantidad 50 mg
Notas de grado de pureza Research
Fórmula molecular C12H23N5O3
Información de solubilidad H2O : 5.2 mg/mL (18.22 mM; ultrasonic and adjust pH to 7 with NaOH) ∣DMSO : 5 mg/mL (17.52 mM; Need ultrasonic)
SMILES N[C@H](C(O)=O)CCCCNC(NCCCC1(N=N1)C)=O
Peso molecular (g/mol) 285.34
Formula Weight (peso de la fórmula) 285.34
Porcentaje de pureza 98.0%